5-Chloro-1-methyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR956907-1G
APOSOR956907-1GEA 806 EUR
APOSOR956907-1G
5-Chloro-1-methyl-1H-pyrazole 95%
5-Chloro-1-methyl-1H-pyrazole
Formel: C₄H₅ClN₂
Molecular Weight: 116,549 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23378582
CAS-Nummer: 42110-76-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR