4-Chloro-1-ethyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR956921-1G
APOSOR956921-1GEA 538 EUR
APOSOR956921-1G
4-Chloro-1-ethyl-1H-pyrazole 95%
4-Chloro-1-ethyl-1H-pyrazole
Formel: C₅H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12026905
CAS-Nummer: 1171076-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR