4-Chloro-1-propyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR956926-1G
APOSOR956926-1GEA 151 EUR
APOSOR956926-1G
4-Chloro-1-propyl-1H-pyrazole 95%
4-Chloro-1-propyl-1H-pyrazole
Formel: C₆H₉ClN₂
Molecular Weight: 144,602 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15976274
CAS-Nummer: 1204901-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR