1-Chloro-2-(methoxymethoxy)benzene 95%

Lieferant: Apollo Scientific

OR956932-1G
APOSOR956932-1GEA 632 EUR
APOSOR956932-1G
1-Chloro-2-(methoxymethoxy)benzene 95%
1-Chloro-2-(methoxymethoxy)benzene
Formel: C₈H₉ClO₂
Molecular Weight: 172,609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21101755
CAS-Nummer: 27701-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR