4-Chloro-3,5-dimethyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR956934-5G
APOSOR956934-5GEA 168 EUR
APOSOR956934-5G
4-Chloro-3,5-dimethyl-1H-pyrazole 95%
4-Chloro-3,5-dimethyl-1H-pyrazole
Formel: C₅H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00825711
CAS-Nummer: 15953-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR