4-Chloro-1,3,5-trimethyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR956935-1G
APOSOR956935-1GEA 538 EUR
APOSOR956935-1G
4-Chloro-1,3,5-trimethyl-1H-pyrazole 95%
4-Chloro-1,3,5-trimethyl-1H-pyrazole
Formel: C₆H₉ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00464340
CAS-Nummer: 56079-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR