2-Butoxy-3-chloropyridine 95%

Lieferant: Apollo Scientific

OR956937-1G
APOSOR956937-1GEA 188 EUR
APOSOR956937-1G
2-Butoxy-3-chloropyridine 95%
2-Butoxy-3-chloropyridine
Formel: C₉H₁₂ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00234558
CAS-Nummer: 100707-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR