5-Chloro-2-(ethylthio)pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-2-(ethylthio)pyridine

OR956943-1G
APOSOR956943-1GEA 632 EUR
APOSOR956943-1G
5-Chloro-2-(ethylthio)pyridine 95%
5-Chloro-2-(ethylthio)pyridine
Formel: C₇H₈ClNS
Molecular Weight: 173,663 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641935
CAS-Nummer: 102645-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR