1-Chloro-2-methoxy-3-methylbenzene 95%
Lieferant: Apollo Scientific
OR956945-1G
APOSOR956945-1GEA
538
EUR
APOSOR956945-1G
1-Chloro-2-methoxy-3-methylbenzene 95%
1-Chloro-2-methoxy-3-methylbenzene
Formel:
C₈H₉ClO Molecular Weight: 156,609 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07780687 CAS-Nummer: 3438-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...