1-Chloro-4-(methoxymethyl)benzene 95%

Lieferant: Apollo Scientific

OR956948-1G
APOSOR956948-1GEA 102 EUR
APOSOR956948-1G
1-Chloro-4-(methoxymethyl)benzene 95%
1-Chloro-4-(methoxymethyl)benzene
Formel: C₈H₉ClO
Molecular Weight: 156,609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22054452
CAS-Nummer: 1195-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR