4-Chloro-1-isopropyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR956950-1G
APOSOR956950-1GEA 364 EUR
APOSOR956950-1G
4-Chloro-1-isopropyl-1H-pyrazole 95%
4-Chloro-1-isopropyl-1H-pyrazole
Formel: C₆H₉ClN₂
Molecular Weight: 144,602 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15976410
CAS-Nummer: 1205921-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR