4-Chloro-1-cyclopropyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR956956-1G
APOSOR956956-1GEA 538 EUR
APOSOR956956-1G
4-Chloro-1-cyclopropyl-1H-pyrazole 95%
4-Chloro-1-cyclopropyl-1H-pyrazole
Formel: C₆H₇ClN₂
Molecular Weight: 142,586 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135674
CAS-Nummer: 1204901-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR