4-Chloro-1-isobutyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR956958-1G
APOSOR956958-1GEA 364 EUR
APOSOR956958-1G
4-Chloro-1-isobutyl-1H-pyrazole 95%
4-Chloro-1-isobutyl-1H-pyrazole
Formel: C₇H₁₁ClN₂
Molecular Weight: 158.629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15976271
CAS-Nummer: 1204901-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR