1-Chloro-3-propoxybenzene 95%
Lieferant: Apollo Scientific
OR956959-1G
APOSOR956959-1GEA
632
EUR
APOSOR956959-1G
1-Chloro-3-propoxybenzene 95%
1-Chloro-3-propoxybenzene
Formel:
C₉H₁₁ClO Molecular Weight: 170,636 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20390265 CAS-Nummer: 51241-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...