1-Chloro-4-propoxy-benzene 95%

Lieferant: Apollo Scientific

OR956960-1G
APOSOR956960-1GEA 632 EUR
APOSOR956960-1G
1-Chloro-4-propoxy-benzene 95%
1-Chloro-4-propoxy-benzene
Formel: C₉H₁₁ClO
Molecular Weight: 170,636 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20390250
CAS-Nummer: 33382-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR