5-Chloro-2-propoxypyridine 95%
Lieferant: Apollo Scientific
OR956961-1G
APOSOR956961-1GEA
632
EUR
APOSOR956961-1G
5-Chloro-2-propoxypyridine 95%
5-Chloro-2-propoxypyridine
Formel:
C₈H₁₀ClNO Molecular Weight: 171,624 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19689764 CAS-Nummer: 1160018-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...