2-Chloro-1-ethoxy-4-methylbenzene 95%

Lieferant: Apollo Scientific

OR956963-1G
APOSOR956963-1GEA 632 EUR
APOSOR956963-1G
2-Chloro-1-ethoxy-4-methylbenzene 95%
2-Chloro-1-ethoxy-4-methylbenzene
Formel: C₉H₁₁ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD23725417
CAS-Nummer: 68758-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR