4-Chloro-1-ethoxy-2-methylbenzene 95%
Lieferant: Apollo Scientific
OR956964-1G
APOSOR956964-1GEA
574
EUR
APOSOR956964-1G
4-Chloro-1-ethoxy-2-methylbenzene 95%
4-Chloro-1-ethoxy-2-methylbenzene
Formel:
C₉H₁₁ClO Molecular Weight: 170,636 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06252403 CAS-Nummer: 35338-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...