3,6-Dichloro-2-methoxypyridine 98%

Lieferant: Apollo Scientific

OR956966-1G
APOSOR956966-1GEA 53.8 EUR
APOSOR956966-1G
3,6-Dichloro-2-methoxypyridine 98%
3,6-Dichloro-2-methoxypyridine
Formel: C₆H₅Cl₂NO
Molecular Weight: 178,016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185545
CAS-Nummer: 1214391-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR