3,5-Dichloro-2-methoxypyridine 95%

Lieferant: Apollo Scientific

OR956967-1G
APOSOR956967-1GEA 538 EUR
APOSOR956967-1G
3,5-Dichloro-2-methoxypyridine 95%
3,5-Dichloro-2-methoxypyridine
Formel: C₆H₅Cl₂NO
Molecular Weight: 178,016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185543
CAS-Nummer: 13472-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR