1-Chloro-4-(methoxymethoxy)-benzene 95%
Lieferant: Apollo Scientific
OR956968-1G
APOSOR956968-1GEA
632
EUR
APOSOR956968-1G
1-Chloro-4-(methoxymethoxy)-benzene 95%
1-Chloro-4-(methoxymethoxy)-benzene
Formel:
C₈H₉ClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD21101771 CAS-Nummer: 826-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...