Butyl[(2-chlorophenyl)methyl]amine 95%

Lieferant: Apollo Scientific

OR956970-1G
APOSOR956970-1GEA 364 EUR
APOSOR956970-1G
Butyl[(2-chlorophenyl)methyl]amine 95%
Butyl[(2-chlorophenyl)methyl]amine
Formel: C₁₁H₁₆ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD07407515
CAS-Nummer: 16183-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR