N-(2-Chlorobenzyl)-N-ethylamine 95%

Lieferant: Apollo Scientific

OR956972-1G
APOSOR956972-1GEA 364 EUR
APOSOR956972-1G
N-(2-Chlorobenzyl)-N-ethylamine 95%
N-(2-Chlorobenzyl)-N-ethylamine
Formel: C₉H₁₂ClN
Molecular Weight: 169,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00045174
CAS-Nummer: 62924-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR