2-Chlorobenzofuran 95%

Lieferant: Apollo Scientific

OR956974-1G
APOSOR956974-1GEA 661 EUR
APOSOR956974-1G
2-Chlorobenzofuran 95%
2-Chlorobenzofuran
Formel: C₈H₅ClO
Molecular Weight: 152,578 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143939
CAS-Nummer: 63361-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR