3-Chlorobenzofuran 95%

Lieferant: Apollo Scientific

OR956976-1G
APOSOR956976-1GEA 806 EUR
APOSOR956976-1G
3-Chlorobenzofuran 95%
3-Chlorobenzofuran
Formel: C₈H₅ClO
Molecular Weight: 152,578 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143940
CAS-Nummer: 63361-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR