1-Butoxy-3-chlorobenzene 95%

Lieferant: Apollo Scientific

OR956983-1G
APOSOR956983-1GEA 632 EUR
APOSOR956983-1G
1-Butoxy-3-chlorobenzene 95%
1-Butoxy-3-chlorobenzene
Formel: C₁₀H₁₃ClO
Molecular Weight: 184,663 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20390445
CAS-Nummer: 51241-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR