5-Chloro-2-dimethylaminopyrimidine 95%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-N,N-dimethylpyrimidin-2-amine

OR956989-1G
APOSOR956989-1GEA 502 EUR
APOSOR956989-1G
5-Chloro-2-dimethylaminopyrimidine 95%
5-Chloro-2-dimethylaminopyrimidine
Formel: C₆H₈ClN₃
Molecular Weight: 157,601 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16657234
CAS-Nummer: 81568-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR