1-Chloro-3-methoxy-5-methylbenzene 95%
Lieferant: Apollo Scientific
OR956991-1G
APOSOR956991-1GEA
538
EUR
APOSOR956991-1G
1-Chloro-3-methoxy-5-methylbenzene 95%
1-Chloro-3-methoxy-5-methylbenzene
Formel:
C₈H₉ClO Molecular Weight: 156,609 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234651 CAS-Nummer: 82477-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...