1-Chloro-3-(2-methoxyethoxy)benzene 95%
Lieferant: Apollo Scientific
OR956992-1G
APOSOR956992-1GEA
632
EUR
APOSOR956992-1G
1-Chloro-3-(2-methoxyethoxy)benzene 95%
1-Chloro-3-(2-methoxyethoxy)benzene
Formel:
C₉H₁₁ClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20390566 CAS-Nummer: 1344249-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...