1-Chloro-4-(2-methoxyethoxy)benzene 95%

Lieferant: Apollo Scientific

OR956993-1G
APOSOR956993-1GEA 632 EUR
APOSOR956993-1G
1-Chloro-4-(2-methoxyethoxy)benzene 95%
1-Chloro-4-(2-methoxyethoxy)benzene
Formel: C₉H₁₁ClO₂
Molecular Weight: 186,635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20390459
CAS-Nummer: 60893-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR