5-Chloro-2-(2-methoxyethoxy)pyridine 95%

Lieferant: Apollo Scientific

OR956994-1G
APOSOR956994-1GEA 632 EUR
APOSOR956994-1G
5-Chloro-2-(2-methoxyethoxy)pyridine 95%
5-Chloro-2-(2-methoxyethoxy)pyridine
Formel: C₈H₁₀ClNO₂
Molecular Weight: 187,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21722841
CAS-Nummer: 1493751-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR