5-Chloro-N-(2-methoxyethyl)pyrimidin-2-amine 95%

Lieferant: Apollo Scientific

OR956997-1G
APOSOR956997-1GEA 0 EUR
APOSOR956997-1G
5-Chloro-N-(2-methoxyethyl)pyrimidin-2-amine 95%
5-Chloro-N-(2-methoxyethyl)pyrimidin-2-amine
Formel: C₇H₁₀ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD23729732
CAS-Nummer: 1506399-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR