4-Chloro-1-cyclopentyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR957002-1G
APOSOR957002-1GEA 451 EUR
APOSOR957002-1G
4-Chloro-1-cyclopentyl-1H-pyrazole 95%
4-Chloro-1-cyclopentyl-1H-pyrazole
Formel: C₈H₁₁ClN₂
Molecular Weight: 170,639 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15976276
CAS-Nummer: 1205839-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR