1-Chloro-3-(propan-2-yloxy)benzene 95%

Lieferant: Apollo Scientific

OR957009-5G
APOSOR957009-5GEA 696 EUR
APOSOR957009-5G
1-Chloro-3-(propan-2-yloxy)benzene 95%
1-Chloro-3-(propan-2-yloxy)benzene
Formel: C₉H₁₁ClO
Molecular Weight: 170,636 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20390264
CAS-Nummer: 51241-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR