5-Chloro-2-(propan-2-ylsulphanyl)pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-2-[(1-methylethyl)thio]pyridine

OR957012-1G
APOSOR957012-1GEA 632 EUR
APOSOR957012-1G
5-Chloro-2-(propan-2-ylsulphanyl)pyridine 95%
5-Chloro-2-(propan-2-ylsulphanyl)pyridine
Formel: C₈H₁₀ClNS
Lagertemperatur: Raumtemperatur
MDL: MFCD21641936
CAS-Nummer: 1489477-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR