2,4-Dichloro-5-methoxypyridine 95%
Lieferant: Apollo Scientific
OR957020-1G
APOSOR957020-1GEA
380
EUR
APOSOR957020-1G
2,4-Dichloro-5-methoxypyridine 95%
2,4-Dichloro-5-methoxypyridine
Formel:
C₆H₅Cl₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD16610792 CAS-Nummer: 1227597-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...