4-Chloro-2,3-dimethoxypyridine 95%

Lieferant: Apollo Scientific

OR957022-1G
APOSOR957022-1GEA 632 EUR
APOSOR957022-1G
4-Chloro-2,3-dimethoxypyridine 95%
4-Chloro-2,3-dimethoxypyridine
Formel: C₇H₈ClNO₂
Molecular Weight: 173,597 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415676
CAS-Nummer: 1261629-46-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR