P-Chlorophenyl methyl sulphoxide 95%

Lieferant: Apollo Scientific

OR957023-1G
APOSOR957023-1GEA 806 EUR
APOSOR957023-1G
P-Chlorophenyl methyl sulphoxide 95%
P-Chlorophenyl methyl sulphoxide
Formel: C₇H₇ClOS
Molecular Weight: 174,648 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040898
CAS-Nummer: 934-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR