1-(4-Chlorophenyl)-n,n-dimethylmethanamine 95%

Lieferant: Apollo Scientific

OR957028-1G
APOSOR957028-1GEA 538 EUR
APOSOR957028-1G
1-(4-Chlorophenyl)-n,n-dimethylmethanamine 95%
1-(4-Chlorophenyl)-n,n-dimethylmethanamine
Formel: C₉H₁₂ClN
Molecular Weight: 169,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00716857
CAS-Nummer: 15184-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR