3-Chloro-4-propoxybenzonitrile 95%

Lieferant: Apollo Scientific

OR957034-1G
APOSOR957034-1GEA 632 EUR
APOSOR957034-1G
3-Chloro-4-propoxybenzonitrile 95%
3-Chloro-4-propoxybenzonitrile
Formel: C₁₀H₁₀ClNO
Molecular Weight: 195.646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14537007
CAS-Nummer: 1216124-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR