1-Chloro-2-(2-methylpropoxy)benzene 95%

Lieferant: Apollo Scientific

OR957037-1G
APOSOR957037-1GEA 632 EUR
APOSOR957037-1G
1-Chloro-2-(2-methylpropoxy)benzene 95%
1-Chloro-2-(2-methylpropoxy)benzene
Formel: C₁₀H₁₃ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD20390248
CAS-Nummer: 60736-65-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR