1-Chloro-3-(2-methylpropoxy)benzene 95%

Lieferant: Apollo Scientific

OR957038-1G
APOSOR957038-1GEA 632 EUR
APOSOR957038-1G
1-Chloro-3-(2-methylpropoxy)benzene 95%
1-Chloro-3-(2-methylpropoxy)benzene
Formel: C₁₀H₁₃ClO
Molecular Weight: 184,663 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20390266
CAS-Nummer: 1343289-00-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR