1-Chloro-4-(2-methylpropoxy)-benzene 95%

Lieferant: Apollo Scientific

OR957039-1G
APOSOR957039-1GEA 632 EUR
APOSOR957039-1G
1-Chloro-4-(2-methylpropoxy)-benzene 95%
1-Chloro-4-(2-methylpropoxy)-benzene
Formel: C₁₀H₁₃ClO
Molecular Weight: 184,663 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20142732
CAS-Nummer: 173416-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR