4-Chloro-2-methyl-1-(propan-2-yloxy)benzene 95%

Lieferant: Apollo Scientific

OR957042-1G
APOSOR957042-1GEA 632 EUR
APOSOR957042-1G
4-Chloro-2-methyl-1-(propan-2-yloxy)benzene 95%
4-Chloro-2-methyl-1-(propan-2-yloxy)benzene
Formel: C₁₀H₁₃ClO
Molecular Weight: 184,663 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23114175
CAS-Nummer: 1369890-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR