3-Chloro-5-methyl-2-(propan-2-yloxy)pyridine 95%

Lieferant: Apollo Scientific

OR957043-1G
APOSOR957043-1GEA 632 EUR
APOSOR957043-1G
3-Chloro-5-methyl-2-(propan-2-yloxy)pyridine 95%
3-Chloro-5-methyl-2-(propan-2-yloxy)pyridine
Formel: C₉H₁₂ClNO
Molecular Weight: 185,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23114143
CAS-Nummer: 1394964-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR