5-Chloro-3-methyl-2-(propan-2-yloxy)pyridine 95%
Lieferant: Apollo Scientific
OR957044-1G
APOSOR957044-1GEA
632
EUR
APOSOR957044-1G
5-Chloro-3-methyl-2-(propan-2-yloxy)pyridine 95%
5-Chloro-3-methyl-2-(propan-2-yloxy)pyridine
Formel:
C₉H₁₂ClNO Molecular Weight: 185,651 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23113730 CAS-Nummer: 1394976-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...