1,4-Dichloro-2-ethoxybenzene 95%

Lieferant: Apollo Scientific

OR957047-1G
APOSOR957047-1GEA 0 EUR
APOSOR957047-1G
1,4-Dichloro-2-ethoxybenzene 95%
1,4-Dichloro-2-ethoxybenzene
Formel: C₈H₈Cl₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD23713614
CAS-Nummer: 84697-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR