3,5-Dichloro-2-ethoxypyridine 95%

Lieferant: Apollo Scientific

OR957048-1G
APOSOR957048-1GEA 632 EUR
APOSOR957048-1G
3,5-Dichloro-2-ethoxypyridine 95%
3,5-Dichloro-2-ethoxypyridine
Formel: C₇H₇Cl₂NO
Molecular Weight: 192,043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06254607
CAS-Nummer: 114755-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR