1-Chloro-2-(cyclopropylmethoxy)benzene 95%

Lieferant: Apollo Scientific

OR957057-1G
APOSOR957057-1GEA 632 EUR
APOSOR957057-1G
1-Chloro-2-(cyclopropylmethoxy)benzene 95%
1-Chloro-2-(cyclopropylmethoxy)benzene
Formel: C₁₀H₁₁ClO
Molecular Weight: 182,647 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17292492
CAS-Nummer: 1281653-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR