5-Chloro-2-(cyclopropylmethoxy)pyridine 95%
Lieferant: Apollo Scientific
OR957061-1G
APOSOR957061-1GEA
632
EUR
APOSOR957061-1G
5-Chloro-2-(cyclopropylmethoxy)pyridine 95%
5-Chloro-2-(cyclopropylmethoxy)pyridine
Formel:
C₉H₁₀ClNO Molecular Weight: 183,635 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21722845 CAS-Nummer: 1489543-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...